Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101731.5 | NP_173308.2 | 2 - 3 | 322 | 116 | |
GenBank mRNA | AK226496.1 | - | 2 - 3 | 160 | 116 | |
AY080701.1 | - | 2 - 3 | 159 | 116 | ||
AY117165.1 | - | 2 - 3 | 100 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |