Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101847.4 | NP_564099.1 | 2 - 3 | 1118 | 131 | |
GenBank mRNA | U06276.1 | - | 2 - 3 | 990 | 131 | |
U40715.1 | - | 2 - 3 | 1010 | 131 | ||
X79210.1 | - | 995 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |