Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101850.2 | NP_564100.1 | 3 - 4 | 429 | 82 | |
GenBank mRNA | AY045614.1 | - | 3 - 4 | 331 | 82 | |
BT001012.1 | - | 2 - 3 | 128 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |