Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101971.3 | NP_173541.1 | 9 - 10 | 1685 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |