Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085336.1 | NP_001078805.1 | 4 - 5 | 725 | 87 | |
NM_126017.5 | NP_201420.1 | 4 - 5 | 730 | 87 | ||
GenBank mRNA | AY072112.1 | - | 4 - 5 | 621 | 87 | |
AY096665.1 | - | 4 - 5 | 543 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |