Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126036.4 | NP_569032.1 | 1 - 2 | 369 | 65 | |
GenBank mRNA | AK227464.1 | - | 1 - 2 | 234 | 65 | |
BT010139.1 | - | 1 - 2 | 102 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |