Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035862.2 | NP_001030939.1 | 5 - 6 | 829 | 71 | |
NM_001035863.1 | NP_001030940.1 | 5 - 6 | 911 | 71 | ||
NM_001035864.1 | NP_001030941.1 | 5 - 6 | 856 | 71 | ||
NM_126206.4 | NP_178254.2 | 5 - 6 | 931 | 71 | ||
GenBank mRNA | AK228266.1 | - | 5 - 6 | 822 | 71 | |
AK317182.1 | - | 5 - 6 | 829 | 71 | ||
BT006469.1 | - | 4 - 5 | 745 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |