Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF73-II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335074.1 | NP_001325256.1 | 9 - 10 | 1167 | 75 | |
NM_126234.3 | NP_178282.2 | 10 - 11 | 1081 | 75 | ||
GenBank mRNA | AK221561.1 | - | 10 - 11 | 1013 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |