Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126363.7 | NP_565294.1 | 7 - 8 | 833 | 90 | |
GenBank mRNA | AY065169.1 | - | 7 - 8 | 795 | 90 | |
AY114571.1 | - | 7 - 8 | 662 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |