Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPBETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111075.3 | NP_186858.1 | 5 - 6 | 1321 | 88 | |
NM_180169.1 | NP_850500.1 | 5 - 6 | 1309 | 88 | ||
GenBank mRNA | AY126990.1 | - | 5 - 6 | 1304 | 88 | |
BT000662.1 | - | 5 - 6 | 1297 | 88 | ||
BT000830.1 | - | 5 - 6 | 1234 | 88 | ||
BT001915.1 | - | 5 - 6 | 1234 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |