Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFB1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337458.1 | NP_001326842.1 | 2 - 3 | 349 | 127 | |
NM_111182.3 | NP_186962.1 | 2 - 3 | 349 | 127 | ||
GenBank mRNA | BT004032.1 | - | 2 - 3 | 306 | 127 | |
BT005084.1 | - | 2 - 3 | 147 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |