Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084632.1 | NP_001078101.1 | 3 - 4 | 550 | 121 | |
NM_001202869.2 | NP_001189798.1 | 6 - 7 | 913 | 121 | ||
NM_111200.5 | NP_566199.4 | 7 - 8 | 1155 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |