Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337465.1 | NP_001327217.1 | 4 - 5 | 667 | 126 | |
NM_001337466.1 | NP_001327218.1 | 4 - 5 | 552 | 126 | ||
NM_111205.4 | NP_566203.2 | 4 - 5 | 552 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |