Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118080.3 | NP_193695.2 | 4 - 5 | 521 | 61 | |
GenBank mRNA | AK117509.1 | - | 4 - 5 | 521 | 61 | |
BT029301.1 | - | 2 - 3 | 311 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |