Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK-HSDH II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118091.3 | NP_193706.2 | 3 - 4 | 764 | 113 | |
NM_202847.3 | NP_974576.1 | 3 - 4 | 805 | 113 | ||
GenBank mRNA | AK228252.1 | - | 3 - 4 | 733 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |