Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203866.1 | NP_001190795.1 | 4 - 5 | 477 | 70 | |
NM_118320.3 | NP_193930.1 | 3 - 4 | 484 | 70 | ||
GenBank mRNA | AF428445.1 | - | 3 - 4 | 436 | 70 | |
AY054175.1 | - | 3 - 4 | 437 | 70 | ||
AY062665.1 | - | 3 - 4 | 428 | 70 | ||
AY093319.1 | - | 3 - 4 | 352 | 70 | ||
AY103313.1 | - | 3 - 4 | 352 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |