Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118352.4 | NP_567658.1 | 4 - 5 | 795 | 73 | |
GenBank mRNA | AF098072.1 | - | 4 - 5 | 710 | 73 | |
AF324663.2 | - | 4 - 5 | 657 | 73 | ||
AF326898.1 | - | 4 - 5 | 657 | 73 | ||
AF339717.1 | - | 4 - 5 | 546 | 73 | ||
AY045699.1 | - | 4 - 5 | 657 | 73 | ||
BT000558.1 | - | 4 - 5 | 546 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |