Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR5-LIKE A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084962.1 | NP_001078431.1 | 7 - 8 | 970 | 106 | |
NM_001084963.1 | NP_001078432.1 | 7 - 8 | 973 | 106 | ||
NM_001341564.1 | NP_001320036.1 | 7 - 8 | 925 | 106 | ||
NM_118418.4 | NP_567672.1 | 7 - 8 | 979 | 106 | ||
NM_179092.1 | NP_849423.1 | 7 - 8 | 979 | 106 | ||
GenBank mRNA | BT000905.1 | - | 7 - 8 | 808 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |