Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203875.2 | NP_001190804.1 | 24 - 25 | 5803 | 113 | |
NM_001341570.1 | NP_001328112.1 | 24 - 25 | 5659 | 113 | ||
NM_118426.4 | NP_194028.2 | 24 - 25 | 5754 | 113 | ||
GenBank mRNA | AY823256.1 | - | 24 - 25 | 5656 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |