Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084964.1 | NP_001078433.1 | 6 - 7 | 1131 | 78 | |
NM_001203877.1 | NP_001190806.1 | 7 - 8 | 1322 | 78 | ||
NM_118430.5 | NP_194032.2 | 6 - 7 | 1160 | 78 | ||
GenBank mRNA | AK227353.1 | - | 6 - 7 | 993 | 78 | |
BT024728.1 | - | 6 - 7 | 809 | 78 | ||
DQ015901.1 | - | 6 - 7 | 860 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |