Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333839.1 | NP_001319269.1 | 3 - 4 | 751 | 106 | |
NM_001333840.1 | NP_001319268.1 | 3 - 4 | 790 | 106 | ||
NM_001333841.1 | NP_001322290.1 | 3 - 4 | 753 | 106 | ||
NM_001333842.1 | NP_001322291.1 | 2 - 3 | 860 | 106 | ||
NM_001333843.1 | NP_001322289.1 | 3 - 4 | 671 | 106 | ||
NM_148599.4 | NP_683440.2 | 2 - 3 | 477 | 106 | ||
GenBank mRNA | KJ920364.1 | - | 2 - 3 | 427 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |