Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102761.4 | NP_174313.1 | 1 - 2 | 288 | 118 | |
GenBank mRNA | AJ973176.1 | - | 1 - 2 | 146 | 118 | |
AY074333.1 | - | 1 - 2 | 178 | 118 | ||
AY123031.1 | - | 1 - 2 | 146 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |