Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102761.4 | NP_174313.1 | 5 - 6 | 1405 | 67 | |
GenBank mRNA | AJ973176.1 | - | 5 - 6 | 1263 | 67 | |
AY074333.1 | - | 5 - 6 | 1295 | 67 | ||
AY123031.1 | - | 5 - 6 | 1263 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |