Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102773.3 | NP_564354.1 | 5 - 6 | 2201 | 69 | |
GenBank mRNA | AY035092.1 | - | 5 - 6 | 1835 | 69 | |
AY063056.1 | - | 5 - 6 | 1791 | 69 | ||
KJ920369.1 | - | 5 - 6 | 1791 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |