Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102785.5 | NP_174337.1 | 15 - 16 | 2454 | 123 | |
GenBank mRNA | AJ440967.1 | - | 15 - 16 | 2105 | 123 | |
AK226570.1 | - | 15 - 16 | 2319 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |