Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSFA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333004.1 | NP_001321152.1 | 1 - 2 | 520 | 121 | |
NM_102966.4 | NP_174511.2 | 1 - 2 | 530 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |