Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198198.1 | NP_001185127.1 | 27 - 28 | 3289 | 65 | |
NM_102984.5 | NP_174527.2 | 27 - 28 | 3314 | 65 | ||
GenBank mRNA | AY074318.1 | - | 27 - 28 | 3237 | 65 | |
AY133872.1 | - | 26 - 27 | 3091 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |