Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204034.2 | NP_001190963.1 | 12 - 13 | 1636 | 65 | |
NM_001342526.1 | NP_001329546.1 | 12 - 13 | 1702 | 65 | ||
NM_120078.2 | NP_195629.2 | 12 - 13 | 2000 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |