Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342528.1 | NP_001329643.1 | 3 - 4 | 955 | 111 | |
NM_120082.4 | NP_195633.1 | 3 - 4 | 693 | 111 | ||
GenBank mRNA | AB010945.1 | - | 3 - 4 | 595 | 111 | |
AY044321.1 | - | 2 - 3 | 534 | 111 | ||
AY086110.1 | - | 3 - 4 | 640 | 111 | ||
BT000929.1 | - | 2 - 3 | 534 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |