Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G16730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127225.3 | NP_179264.2 | 12 - 13 | 1721 | 102 | |
GenBank mRNA | AJ270309.1 | - | 12 - 13 | 1542 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |