Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127281.2 | NP_179318.1 | 1 | 1232 | 107 | |
GenBank mRNA | AY128313.1 | - | 1 | 1199 | 107 | |
BT001174.1 | - | 1 | 1099 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |