Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127294.5 | NP_179331.1 | 8 - 9 | 1092 | 81 | |
GenBank mRNA | BT024730.1 | - | 7 - 8 | 931 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |