Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036287.3 | NP_001031364.2 | 3 - 4 | 328 | 106 | |
NM_001084440.1 | NP_001077909.1 | 4 - 5 | 256 | 106 | ||
NM_127310.3 | NP_179347.1 | 4 - 5 | 358 | 106 | ||
GenBank mRNA | AF370562.1 | - | 3 - 4 | 191 | 106 | |
AK317044.1 | - | 4 - 5 | 227 | 106 | ||
AY150467.1 | - | 3 - 4 | 191 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |