Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPDHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036448.2 | NP_001031525.1 | 3 - 4 | 918 | 92 | |
NM_001336924.1 | NP_001325243.1 | 1 - 2 | 1612 | 92 | ||
NM_129717.5 | NP_181685.1 | 3 - 4 | 1155 | 92 | ||
NM_180021.5 | NP_850352.1 | 2 - 3 | 1224 | 92 | ||
GenBank mRNA | AJ347019.1 | - | 3 - 4 | 812 | 92 | |
AY063920.1 | - | 2 - 3 | 1066 | 92 | ||
AY091247.1 | - | 1 - 2 | 657 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |