Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180062.3 | NP_850393.1 | 1 - 2 | 464 | 86 | |
GenBank mRNA | AJ973175.1 | - | 1 - 2 | 151 | 86 | |
AY063901.1 | - | 1 - 2 | 432 | 86 | ||
AY096505.1 | - | 1 - 2 | 151 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |