Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337221.1 | NP_001318437.1 | 7 - 8 | 2066 | 84 | |
NM_180129.4 | NP_850460.1 | 7 - 8 | 1792 | 84 | ||
NM_180130.2 | NP_850461.1 | 4 - 5 | 1320 | 84 | ||
GenBank mRNA | AY519511.1 | - | 6 - 7 | 1462 | 84 | |
BT001096.1 | - | 3 - 4 | 1216 | 84 | ||
BT001105.1 | - | 3 - 4 | 1216 | 84 | ||
U28422.1 | - | 7 - 8 | 1699 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |