Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125092.1 | NP_001118564.1 | 7 - 8 | 1414 | 60 | |
NM_111249.5 | NP_187028.1 | 8 - 9 | 1568 | 60 | ||
NM_180176.4 | NP_850507.1 | 8 - 9 | 1664 | 60 | ||
GenBank mRNA | AJ608674.1 | - | 7 - 8 | 1371 | 60 | |
AY084559.1 | - | 8 - 9 | 1445 | 60 | ||
AY150385.1 | - | 7 - 8 | 1366 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |