Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111458.5 | NP_187235.1 | 4 - 5 | 748 | 101 | |
NM_180189.4 | NP_850520.1 | 4 - 5 | 476 | 101 | ||
GenBank mRNA | AK227034.1 | - | 4 - 5 | 516 | 101 | |
AK316673.1 | - | 4 - 5 | 545 | 101 | ||
AY062713.1 | - | 4 - 5 | 461 | 101 | ||
AY093347.1 | - | 3 - 4 | 270 | 101 | ||
X75431.1 | - | 443 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |