Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202993.1 | NP_001189922.1 | 7 - 8 | 1630 | 109 | |
NM_001338329.1 | NP_001327710.1 | 6 - 7 | 1767 | 109 | ||
NM_112692.3 | NP_566597.1 | 5 - 6 | 1735 | 109 | ||
NM_180276.2 | NP_850607.1 | 6 - 7 | 1665 | 109 | ||
GenBank mRNA | AY033827.1 | - | 5 - 6 | 1609 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |