Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342547.1 | NP_001328757.1 | 3 - 4 | 861 | 143 | |
NM_120126.5 | NP_568064.1 | 3 - 4 | 806 | 143 | ||
GenBank mRNA | AK227118.1 | - | 3 - 4 | 668 | 143 | |
AY054264.1 | - | 3 - 4 | 660 | 143 | ||
BT002306.1 | - | 3 - 4 | 636 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |