Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342579.1 | NP_001331107.1 | 2 - 3 | 592 | 121 | |
NM_120200.4 | NP_568085.2 | 2 - 3 | 646 | 121 | ||
GenBank mRNA | AF454354.1 | - | 2 - 3 | 414 | 121 | |
BT005796.1 | - | 2 - 3 | 414 | 121 | ||
KJ138712.1 | - | 2 - 3 | 414 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |