Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342617.1 | NP_001330425.1 | 4 - 5 | 864 | 77 | |
NM_001342618.1 | NP_001330424.1 | 4 - 5 | 864 | 77 | ||
NM_001342619.1 | NP_001330427.1 | 4 - 5 | 833 | 77 | ||
NM_001342620.1 | NP_001330426.1 | 4 - 5 | 833 | 77 | ||
NM_120255.3 | NP_195797.3 | 4 - 5 | 833 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |