Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120404.3 | NP_195946.2 | 4 - 5 | 709 | 66 | |
GenBank mRNA | AY065128.1 | - | 4 - 5 | 709 | 66 | |
AY081592.1 | - | 4 - 5 | 623 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |