Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342706.1 | NP_001331919.1 | 1 - 2 | 165 | 78 | |
NM_120425.3 | NP_568119.1 | 1 - 2 | 165 | 78 | ||
GenBank mRNA | AK117898.1 | - | 1 - 2 | 142 | 78 | |
BT003658.1 | - | 1 - 2 | 129 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |