Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120454.4 | NP_195993.1 | 8 - 9 | 1977 | 84 | |
NM_180429.4 | NP_850760.1 | 8 - 9 | 1977 | 84 | ||
GenBank mRNA | AK227171.1 | - | 8 - 9 | 1986 | 84 | |
L08789.1 | - | 8 - 9 | 1905 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |