Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335703.1 | NP_001324426.1 | 1 - 2 | 522 | 69 | |
NM_127640.3 | NP_179668.1 | 1 - 2 | 522 | 69 | ||
GenBank mRNA | AY084478.1 | - | 1 - 2 | 234 | 69 | |
BT003932.1 | - | 1 - 2 | 232 | 69 | ||
BT005024.1 | - | 1 - 2 | 170 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |