Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127645.4 | NP_179673.1 | 3 - 4 | 1185 | 71 | |
GenBank mRNA | DQ446535.1 | - | 3 - 4 | 1053 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |