Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335713.1 | NP_001324598.1 | 1 - 2 | 371 | 68 | |
NM_127654.4 | NP_565489.2 | 1 - 2 | 371 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |