Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127659.4 | NP_565491.1 | 3 - 4 | 866 | 78 | |
GenBank mRNA | AY062799.1 | - | 3 - 4 | 777 | 78 | |
AY081596.1 | - | 3 - 4 | 667 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |