Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339583.1 | NP_001319732.1 | 4 - 5 | 786 | 77 | |
NM_001339584.1 | NP_001326182.1 | 2 - 3 | 558 | 77 | ||
NM_115132.3 | NP_566971.2 | 3 - 4 | 1328 | 77 | ||
NM_180354.2 | NP_850685.1 | 3 - 4 | 452 | 77 | ||
GenBank mRNA | U73462.1 | - | 3 - 4 | 452 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |